ENA375228299 Enamine

6,6-dimethyl-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carboxylic acid

  • MDL number MFCD20319876
  • Cas Number : 1342675-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114622

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114622
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart