ENA375228319 Enamine

3-[3-(benzyloxy)phenyl]-3-hydroxy-2,2-dimethylpropanoic acid

  • MDL number MFCD22566088
  • Cas Number : 1423028-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116649
Availability details for
Quantity
  • Availability
  • Add to Cart