ENA375228323 Enamine

2-chloro-1-(4-phenyl-1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one

  • MDL number MFCD22566093
  • Cas Number : 1423033-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47567

Safety Information

msds https://www.enaminestore.com/catalog/EN300-47567
Availability details for
Quantity
  • Availability
  • Add to Cart