ENA375228326 Enamine

4-(propan-2-yl)cycloheptan-1-one

  • MDL number MFCD22566095
  • Cas Number : 13656-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85706
Availability details for
Quantity
  • Availability
  • Add to Cart