ENA375228333 Enamine

3-(propan-2-yl)-1,3,7-triazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD22566103
  • Cas Number : 1423026-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94144
Availability details for
Quantity
  • Availability
  • Add to Cart