ENA375228346 Enamine

1-(2-chloroethyl)-5-nitro-1,2-dihydropyridin-2-one

  • MDL number MFCD11164695
  • Cas Number : 154011-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40161
Availability details for
Quantity
  • Availability
  • Add to Cart