ENA375228352 Enamine

2-chloro-N-(2-methylpropyl)-N-(propan-2-yl)acetamide

  • MDL number MFCD12798905
  • Cas Number : 39096-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47519
Availability details for
Quantity
  • Availability
  • Add to Cart