ENA375228366 Enamine

2-(3-nitrophenyl)oxirane

  • MDL number MFCD10699484
  • Cas Number : 20697-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61700
Availability details for
Quantity
  • Availability
  • Add to Cart