ENA375228371 Enamine

1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00464254
  • Cas Number : 25016-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66976

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-66976
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart