ENA375228391 Enamine

4,6-dichloro-N,N,5-trimethylpyrimidin-2-amine

  • MDL number MFCD18844077
  • Cas Number : 61770-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116598
Availability details for
Quantity
  • Availability
  • Add to Cart