ENA375228395 Enamine

oxolane-2-carbonitrile

  • MDL number MFCD07787008
  • Cas Number : 14631-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88010

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-88010
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart