ENA375228396 Enamine

5-(chloromethyl)-2-methoxybenzonitrile

  • MDL number MFCD18394249
  • Cas Number : 109418-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88601

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88601
Availability details for
Quantity
  • Availability
  • Add to Cart