ENA375228406 Enamine

4-(2-hydroxypropan-2-yl)benzonitrile

  • MDL number MFCD00096234
  • Cas Number : 77802-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116705
Availability details for
Quantity
  • Availability
  • Add to Cart