ENA375228408 Enamine

potassium trifluoro[4-(oxan-4-yloxy)phenyl]boranuide

  • MDL number MFCD19059903

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116723
Availability details for
Quantity
  • Availability
  • Add to Cart