ENA379363388 Enamine

3-amino-2-methyl-1lambda6-benzothiophene-1,1-dione

  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1,1-dioxido-1-benzothien-3-ylamine
Inchi Code 1S/C9H9NO2S/c1-6-9(10)7-4-2-3-5-8(7)13(6,11)12/h2-5H,10H2,1H3
Inchi Key NPOTZCAVWDMPNM-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112875

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-112875
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart