ENA379363400 Enamine

4-methoxy-3,4-dihydro-2H-1-benzopyran-8-amine

  • MDL number MFCD22578467
  • Cas Number : 1423031-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113565
Availability details for
Quantity
  • Availability
  • Add to Cart