ENA379363401 Enamine

N4,N4-dimethyl-3,4-dihydro-2H-1-benzopyran-4,8-diamine

  • MDL number MFCD22578468
  • Cas Number : 1423025-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113566
Availability details for
Quantity
  • Availability
  • Add to Cart