ENA379363403 Enamine

2-bromo-2-propylpentanoic acid

  • MDL number MFCD22578470
  • Cas Number : 98551-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113660

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-113660
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart