ENA379363414 Enamine

sodium 2-(2,3-dihydro-1H-indol-1-yl)propanoate

  • MDL number MFCD22578482
  • Cas Number : 1423024-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115612
Availability details for
Quantity
  • Availability
  • Add to Cart