ENA379363416 Enamine

3,5-diethyl-1-methyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD22578484
  • Cas Number : 1423024-95-6
  • Molecular Weight : 217.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115660
Availability details for
Quantity
  • Availability
  • Add to Cart