ENA379363419 Enamine

2-(1-benzyl-1H-pyrazol-4-yl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD22578487
  • Cas Number : 1423028-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115973
Availability details for
Quantity
  • Availability
  • Add to Cart