ENA379363430 Enamine

1-(2,3-dihydro-1-benzofuran-3-yl)ethan-1-amine

  • MDL number MFCD22578498
  • Cas Number : 1423025-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116047
Availability details for
Quantity
  • Availability
  • Add to Cart