ENA379363439 Enamine

methyl 2-[3-(aminomethyl)piperidin-1-yl]acetate dihydrochloride

  • MDL number MFCD22578510
  • Cas Number : 1423024-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116678
Availability details for
Quantity
  • Availability
  • Add to Cart