ENA379363443 Enamine

2-(5-amino-3-tert-butyl-1H-pyrazol-1-yl)acetic acid hydrochloride

  • MDL number MFCD22578514
  • Cas Number : 1423033-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116709
Availability details for
Quantity
  • Availability
  • Add to Cart