ENA379363444 Enamine

4-(chlorosulfonyl)-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD22578515
  • Cas Number : 1423031-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116718

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-116718
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart