ENA379363456 Enamine

(2,4,5-trifluorophenyl)hydrazine hydrochloride

  • MDL number MFCD22578527
  • Cas Number : 1024006-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116841
Availability details for
Quantity
  • Availability
  • Add to Cart