ENA379363463 Enamine

3-amino-N-[1-(5-chlorothiophen-2-yl)ethyl]-3-phenylpropanamide hydrochloride

  • MDL number MFCD22578534
  • Cas Number : 1423029-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116924

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116924
Availability details for
Quantity
  • Availability
  • Add to Cart