ENA379363466 Enamine

N-(2-amino-2-phenylethyl)-2,5-dimethylthiophene-3-sulfonamide hydrochloride

  • MDL number MFCD22578537
  • Cas Number : 1423034-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-116927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116927
Availability details for
Quantity
  • Availability
  • Add to Cart