ENA379363469 Enamine

2-amino-N-benzyl-4-methyl-N-propylpentanamide hydrochloride

  • MDL number MFCD22578540
  • Cas Number : 1423028-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116930

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116930
Availability details for
Quantity
  • Availability
  • Add to Cart