ENA379363471 Enamine

2-(3-cycloheptyl-1,2,4-oxadiazol-5-yl)azepane hydrochloride

  • MDL number MFCD22578542
  • Cas Number : 1423026-03-2
  • Molecular Weight : 299.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116966
Availability details for
Quantity
  • Availability
  • Add to Cart