ENA379363482 Enamine

2-(methylamino)-2-(pyridin-4-yl)propanamide dihydrochloride

  • MDL number MFCD22578554
  • Cas Number : 1423029-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117035
Availability details for
Quantity
  • Availability
  • Add to Cart