ENA379363489 Enamine

(2S)-2-amino-N-(2-methoxyethyl)-4-methylpentanamide hydrochloride

  • MDL number MFCD22578561
  • Cas Number : 1423040-71-4
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-117069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117069
Availability details for
Quantity
  • Availability
  • Add to Cart