ENA379363492 Enamine

methyl 3-(1-chloro-2-methoxy-2-oxoethyl)benzoate

  • MDL number MFCD22578565
  • Cas Number : 1423026-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117111
Availability details for
Quantity
  • Availability
  • Add to Cart