ENA379363497 Enamine

2-(3,6-dihydro-2H-pyran-4-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22578571
  • Cas Number : 1423026-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117153

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117153
Availability details for
Quantity
  • Availability
  • Add to Cart