ENA379363500 Enamine

2-bromo-1-(4-methylpiperazin-1-yl)ethan-1-one hydrobromide

  • MDL number MFCD22578574
  • Cas Number : 1423029-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117159

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-117159
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart