ENA379363520 Enamine

(1S)-7-(propan-2-yloxy)-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD22578597
  • Cas Number : 1423040-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117402
Availability details for
Quantity
  • Availability
  • Add to Cart