ENA379363522 Enamine

1-{5-acetyl-4H,5H,6H,7H-thieno[3,2-c]pyridin-2-yl}-2-chloroethan-1-one

  • MDL number MFCD22578599
  • Cas Number : 1423032-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117408
Availability details for
Quantity
  • Availability
  • Add to Cart