ENA379363530 Enamine

methyl 2-[4-(aminomethyl)-3,5-dimethyl-1H-pyrazol-1-yl]acetate dihydrochloride

  • MDL number MFCD22578607
  • Cas Number : 1423034-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117499
Availability details for
Quantity
  • Availability
  • Add to Cart