ENA379363533 Enamine

N-(4-aminobutyl)-2-chloroacetamide hydrochloride

  • MDL number MFCD22578610
  • Cas Number : 1423029-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117544
Availability details for
Quantity
  • Availability
  • Add to Cart