ENA379363536 Enamine

1,4-dimethyl-1,4-diazepane-2-carbonitrile

  • MDL number MFCD22578613
  • Cas Number : 1423032-36-3
  • Molecular Weight : 153.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,4-dimethyl-1,4-diazepane-2-carbonitrile
Inchi Code 1S/C8H15N3/c1-10-4-3-5-11(2)8(6-9)7-10/h8H,3-5,7H2,1-2H3
Inchi Key YYISHFQLPFEQOM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-117551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117551
Availability details for
Quantity
  • Availability
  • Add to Cart