ENA379363539 Enamine

N-(2-amino-2-phenylethyl)thiophene-2-sulfonamide hydrochloride

  • MDL number MFCD22578616
  • Cas Number : 1423033-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-117579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117579
Availability details for
Quantity
  • Availability
  • Add to Cart