ENA379363548 Enamine

3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid hydrochloride

  • MDL number MFCD22578629
  • Cas Number : 1423032-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117746
Availability details for
Quantity
  • Availability
  • Add to Cart