ENA379363553 Enamine

[1-(4-fluorophenyl)-3-methylazetidin-3-yl]methanol

  • MDL number MFCD22578634
  • Cas Number : 1423026-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117766

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117766
Availability details for
Quantity
  • Availability
  • Add to Cart