ENA379363554 Enamine

1-(2,2,2-trifluoroethyl)piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD22578635
  • Cas Number : 50586-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117770
Availability details for
Quantity
  • Availability
  • Add to Cart