ENA379363559 Enamine

3-amino-1-(2,6-dimethylmorpholin-4-yl)-3-phenylpropan-1-one hydrochloride

  • MDL number MFCD22578641
  • Cas Number : 1423032-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117856
Availability details for
Quantity
  • Availability
  • Add to Cart