ENA379363560 Enamine

2-{7-methylimidazo[1,2-a]pyrimidin-2-yl}ethan-1-amine dihydrobromide

  • MDL number MFCD22578642
  • Cas Number : 1423032-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117875
Availability details for
Quantity
  • Availability
  • Add to Cart