ENA379363563 Enamine

N-(2-fluorophenyl)-2-[(prop-2-yn-1-yl)amino]acetamide hydrochloride

  • MDL number MFCD22578645
  • Cas Number : 1423031-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117880
Availability details for
Quantity
  • Availability
  • Add to Cart