ENA379363564 Enamine

4-(chloromethyl)-5-(thiophen-2-yl)-1,2-oxazole

  • MDL number MFCD22578646
  • Cas Number : 1423031-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117942
Availability details for
Quantity
  • Availability
  • Add to Cart