ENA379363568 Enamine

2-(propan-2-yloxy)ethanimidamide hydrochloride

  • MDL number MFCD22578651
  • Cas Number : 1423033-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117966
Availability details for
Quantity
  • Availability
  • Add to Cart