ENA379363571 Enamine

2-amino-N-methyl-N-(pyridin-2-ylmethyl)butanamide dihydrochloride

  • MDL number MFCD22578654
  • Cas Number : 1423024-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118011
Availability details for
Quantity
  • Availability
  • Add to Cart